Product Details

SNP ID
rs144302745
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:98666984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTTCATGTGAAGATGTCTGTA[C/T]GATTTCAGCCTCCAGCATTCATGTG
Phenotype
MIM: 614904
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNX7 PubMed Links

Gene Details

Gene
SNX7
Gene Name
sorting nexin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015976.4 Intron NP_057060.2
NM_152238.3 Intron NP_689424.2
XM_006710677.2 Intron XP_006710740.1
XM_006710678.2 Intron XP_006710741.1
XM_011541564.2 Intron XP_011539866.1
XM_017001425.1 Intron XP_016856914.1
XM_017001426.1 Intron XP_016856915.1
XM_017001427.1 Intron XP_016856916.1
XM_017001428.1 Intron XP_016856917.1

View Full Product Details