Product Details

SNP ID
rs144335029
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:1449746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCAATGGGCCACCGCGCCCGG[C/T]CAGAAGATTTTTATGGTAAAATTTT
Phenotype
MIM: 611901
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATAD3C PubMed Links

Gene Details

Gene
ATAD3C
Gene Name
ATPase family, AAA domain containing 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039211.2 58 UTR 5 NP_001034300.2
Gene
VWA1
Gene Name
von Willebrand factor A domain containing 1
There are no transcripts associated with this gene.

View Full Product Details