Product Details

SNP ID
rs144420834
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:108136792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCCTTCTTTGGAAATAATTCGT[A/C]GAAAGAGGCCAACCATATTCAGCTG
Phenotype
MIM: 608744
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A24 PubMed Links

Gene Details

Gene
SLC25A24
Gene Name
solute carrier family 25 member 24
There are no transcripts associated with this gene.

View Full Product Details