Product Details

SNP ID
rs144501580
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:43451957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCTGCTGGGGCGTGTCCAGGAA[A/G]TACTGGGGGTCAGTGATGCGGGTGT
Phenotype
MIM: 615463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HYI PubMed Links

Gene Details

Gene
HYI
Gene Name
hydroxypyruvate isomerase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190880.2 1006 Silent Mutation TAC,TAT Y161Y NP_001177809.1
NM_001243526.1 1006 Silent Mutation TAC,TAT Y186Y NP_001230455.1
NM_031207.5 1006 Silent Mutation TAC,TAT Y161Y NP_112484.3
XM_005271239.4 1006 Silent Mutation TAC,TAT Y186Y XP_005271296.1
XM_005271240.4 1006 Silent Mutation TAC,TAT Y152Y XP_005271297.1
XM_006710937.3 1006 Silent Mutation TAC,TAT Y186Y XP_006711000.1
XM_011542222.2 1006 Silent Mutation TAC,TAT Y152Y XP_011540524.1
Gene
MIR6735
Gene Name
microRNA 6735
There are no transcripts associated with this gene.

Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015284.3 1006 UTR 3 NP_056099.3
XM_005270686.3 1006 Intron XP_005270743.1
XM_006710501.3 1006 Intron XP_006710564.1
XM_011541106.2 1006 Intron XP_011539408.1
XM_011541107.2 1006 Intron XP_011539409.1
XM_017000819.1 1006 Intron XP_016856308.1
XM_017000820.1 1006 Intron XP_016856309.1
XM_017000821.1 1006 Intron XP_016856310.1

View Full Product Details