Product Details
- SNP ID
-
rs144947476
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:158398733 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCATAGTGGACAAAGACCACAGTG[A/C]GATGTGAGGCACAGGTGACAAAGGC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10T2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34465440] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10T2
- Gene Name
- olfactory receptor family 10 subfamily T member 2
View Full Product Details