Product Details

SNP ID
rs145188643
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:181089307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTTTTTCAGGGCGGAGAGGCGGA[C/T]GCGACGGAAGCTGCCTGGAGCCGCG
Phenotype
MIM: 607177
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IER5 PubMed Links

Gene Details

Gene
IER5
Gene Name
immediate early response 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016545.4 806 Silent Mutation GAC,GAT D135D NP_057629.2

View Full Product Details