Product Details

SNP ID
rs145828311
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:200407399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCTCTAAGTTCTTCTGAGAATC[C/T]ATCAGATCATCCAGCTCCTGGGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF281 PubMed Links

Gene Details

Gene
ZNF281
Gene Name
zinc finger protein 281
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281293.1 3819 Missense Mutation ATA,ATG I769M NP_001268222.1
NM_001281294.1 3819 Missense Mutation ATA,ATG I733M NP_001268223.1
NM_012482.4 3819 Missense Mutation ATA,ATG I769M NP_036614.1
XM_017000887.1 3819 Missense Mutation ATA,ATG I769M XP_016856376.1
XM_017000888.1 3819 Missense Mutation ATA,ATG I769M XP_016856377.1

View Full Product Details