Product Details
- SNP ID
-
rs145916199
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:155868138 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCAGCATATACCCAGAGACCCTC[A/G]GCAACAAGAAGAAAATCATCAAAGT
- Phenotype
-
MIM: 610393
MIM: 608741
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GON4L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs822522] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GON4L
- Gene Name
- gon-4 like
There are no transcripts associated with this gene.
- Gene
- SYT11
- Gene Name
- synaptotagmin 11
View Full Product Details