Product Details

SNP ID
rs145943125
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:240493221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGATGGTGCGGCTCCGGCAGCC[C/T]TCCTCGCTCACCGTCTGCCGCAGCG
Phenotype
MIM: 608832
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GREM2 PubMed Links

Gene Details

Gene
GREM2
Gene Name
gremlin 2, DAN family BMP antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022469.3 535 Silent Mutation GAA,GAG E85E NP_071914.3
XM_005273226.3 535 Silent Mutation GAA,GAG E103E XP_005273283.1
XM_011544249.2 535 Silent Mutation GAA,GAG E85E XP_011542551.1

View Full Product Details