Product Details

SNP ID
rs146139173
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:53852147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTAAACGGGTGATTAGGCGCCA[A/G]TGTGGCACCATCCAGACACTGGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YIPF1 PubMed Links

Gene Details

Gene
YIPF1
Gene Name
Yip1 domain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018982.4 1182 UTR 3 NP_061855.1
XM_017001499.1 1182 UTR 3 XP_016856988.1
XM_017001500.1 1182 UTR 3 XP_016856989.1
XM_017001501.1 1182 UTR 3 XP_016856990.1

View Full Product Details