Product Details

SNP ID
rs146195432
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:180956929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAATCTGCCACTAACTGGCACC[A/G]CTTTTGTGACCCCTCTCCCCACTCC
Phenotype
MIM: 603944
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STX6 PubMed Links

Gene Details

Gene
STX6
Gene Name
syntaxin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286210.1 Intron NP_001273139.1
NM_005819.5 Intron NP_005810.1
XM_011509058.1 Intron XP_011507360.1
XM_011509059.1 Intron XP_011507361.1
XM_017000006.1 Intron XP_016855495.1
XM_017000007.1 Intron XP_016855496.1

View Full Product Details