Product Details

SNP ID
rs146416790
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:159873139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCCTTTTTCTCCTCCTCCAGC[C/T]GCTCCTTCTCAATCTGTTCTCTCTG
Phenotype
MIM: 605152
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP45 PubMed Links

Gene Details

Gene
CFAP45
Gene Name
cilia and flagella associated protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012337.2 1400 Missense Mutation CAG,CGG Q461R NP_036469.2

View Full Product Details