Product Details

SNP ID
rs146627411
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:247965854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTCTATCAATGCTTTCATACAT[A/G]CATTGTATGTGTTTCAGCTTCCATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2AK2 PubMed Links

Gene Details

Gene
OR2AK2
Gene Name
olfactory receptor family 2 subfamily AK member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004491.1 523 Missense Mutation ACA,GCA T175A NP_001004491.1
Gene
OR2L13
Gene Name
olfactory receptor family 2 subfamily L member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304535.1 523 Intron NP_001291464.1
NM_175911.3 523 Intron NP_787107.1
XM_011544169.2 523 Intron XP_011542471.1

View Full Product Details