Product Details

SNP ID
rs146674823
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:205599146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGGCAGATATTCCAGGCTCAG[A/G]GCAGCTATAGTTTCCTGGTCTGTGG
Phenotype
MIM: 600246
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELK4 PubMed Links

Gene Details

Gene
ELK4
Gene Name
ELK4, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001973.3 1470 Intron NP_001964.2
NM_021795.2 1470 Intron NP_068567.1
XM_005244950.4 1470 Intron XP_005245007.1
XM_005244951.4 1470 Intron XP_005245008.1
XM_017000580.1 1470 Intron XP_016856069.1
Gene
MFSD4A
Gene Name
major facilitator superfamily domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181644.4 1470 Missense Mutation AGC,GGC S462G NP_857595.3
XM_011509229.2 1470 Missense Mutation AGC,GGC S375G XP_011507531.1

View Full Product Details