Product Details

SNP ID
rs146709942
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:109250665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACAGTGCCGGAGAACCAGCCA[C/G]CAGGCACCCCTGTTGCATCCCTGAG
Phenotype
MIM: 604265
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CELSR2 PubMed Links

Gene Details

Gene
CELSR2
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001408.2 647 Missense Mutation CCA,GCA P196A NP_001399.1
XM_005270580.4 647 Missense Mutation CCA,GCA P196A XP_005270637.1

View Full Product Details