Product Details
- SNP ID
-
rs146749656
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:77697892 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTAAGCATCACATTACCACATTTA[C/G]TGACTGCAATCTTAGTATTGTCAAT
- Phenotype
-
MIM: 615146
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
USP33
PubMed Links
Gene Details
- Gene
- USP33
- Gene Name
- ubiquitin specific peptidase 33
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015017.4 |
3016 |
Missense Mutation |
ACT,AGT |
T881S |
NP_055832.3 |
NM_201624.2 |
3016 |
Missense Mutation |
ACT,AGT |
T850S |
NP_963918.1 |
NM_201626.2 |
3016 |
Intron |
|
|
NP_963920.1 |
XM_005270648.1 |
3016 |
Missense Mutation |
ACT,AGT |
T873S |
XP_005270705.1 |
XM_005270649.1 |
3016 |
Missense Mutation |
CTA,GTA |
L847V |
XP_005270706.1 |
XM_011541055.1 |
3016 |
Missense Mutation |
ACT,AGT |
T782S |
XP_011539357.1 |
XM_011541056.2 |
3016 |
Intron |
|
|
XP_011539358.1 |
XM_017000722.1 |
3016 |
Missense Mutation |
ACT,AGT |
T842S |
XP_016856211.1 |
XM_017000723.1 |
3016 |
Missense Mutation |
CTA,GTA |
L816V |
XP_016856212.1 |
XM_017000724.1 |
3016 |
Missense Mutation |
CTA,GTA |
L808V |
XP_016856213.1 |
XM_017000725.1 |
3016 |
Missense Mutation |
ACT,AGT |
T751S |
XP_016856214.1 |
XM_017000726.1 |
3016 |
Missense Mutation |
ACT,AGT |
T743S |
XP_016856215.1 |
XM_017000727.1 |
3016 |
Missense Mutation |
ACT,AGT |
T702S |
XP_016856216.1 |
XM_017000728.1 |
3016 |
Missense Mutation |
ACT,AGT |
T702S |
XP_016856217.1 |
XM_017000729.1 |
3016 |
Missense Mutation |
CTA,GTA |
L748V |
XP_016856218.1 |
View Full Product Details