Product Details

SNP ID
rs146859968
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:110517625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAAGTAGACTGCACTGGAGAAG[A/G]GGATGACTCCAATGAAGAGAAAGAA
Phenotype
MIM: 602420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNA10 PubMed Links

Gene Details

Gene
KCNA10
Gene Name
potassium voltage-gated channel subfamily A member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005549.2 1551 Missense Mutation CCC,CTC P388L NP_005540.1

View Full Product Details