Product Details

SNP ID
rs146875514
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:152984631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGTAGATAGTAATTGGCATGA[A/C]GGAGATGGGCAGAGAAGGGCTGTGC
Phenotype
MIM: 182265
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPRR1A PubMed Links

Gene Details

Gene
SPRR1A
Gene Name
small proline rich protein 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199828.1 Intron NP_001186757.1
NM_005987.3 Intron NP_005978.2

View Full Product Details