Product Details

SNP ID
rs147087321
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:68046904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCTTTGATCTTGCAGATCAGCT[C/T]ATAGAAGGCCTTCAGCTCTTCCAGG
Phenotype
MIM: 605193 MIM: 615406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIRAS3 PubMed Links

Gene Details

Gene
DIRAS3
Gene Name
DIRAS family GTPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004675.3 1674 Missense Mutation AAG,GAG K132E NP_004666.1
XM_017002773.1 1674 Missense Mutation AAG,GAG K132E XP_016858262.1
Gene
GNG12-AS1
Gene Name
GNG12 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details