Product Details

SNP ID
rs147127811
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:94171733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGCCCTAAAGGCAGAAAGGAC[C/T]CAAAAGTGAAATTCTCATTTTTAAG
Phenotype
MIM: 610496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP29 PubMed Links

Gene Details

Gene
ARHGAP29
Gene Name
Rho GTPase activating protein 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004815.3 6291 UTR 3 NP_004806.3
XM_006711048.3 6291 Intron XP_006711111.1
XM_011542438.2 6291 Intron XP_011540740.1
XM_011542439.2 6291 Intron XP_011540741.1
XM_011542440.2 6291 Intron XP_011540742.1

View Full Product Details