Product Details

SNP ID
rs147784232
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:212044715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATGAAGAAGGCTTTGTTCGATT[G/T]TATAACACAGAATCACAAAGTTTCA
Phenotype
MIM: 610617 MIM: 611350
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DTL PubMed Links

Gene Details

Gene
DTL
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286229.1 446 UTR 5 NP_001273158.1
NM_001286230.1 446 Silent Mutation TTG,TTT L36F NP_001273159.1
NM_016448.3 446 Silent Mutation TTG,TTT L78F NP_057532.3
XM_011509614.1 446 Missense Mutation TTG,TTT L16F XP_011507916.1
Gene
INTS7
Gene Name
integrator complex subunit 7
There are no transcripts associated with this gene.

Gene
RPL21P28
Gene Name
ribosomal protein L21 pseudogene 28
There are no transcripts associated with this gene.

View Full Product Details