Product Details

SNP ID
rs148309294
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:181088948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCGCATCGTCAGCATCTCTCTG[A/G]GCAAGATCTACAACTCGCGGGTCCA
Phenotype
MIM: 607177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IER5 PubMed Links

Gene Details

Gene
IER5
Gene Name
immediate early response 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016545.4 447 Missense Mutation AGC,GGC S16G NP_057629.2

View Full Product Details