Product Details

SNP ID
rs148806389
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:20140218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGTGGCCATCCTTGCTGGCAGC[A/G]GTGAGTAAAGACTCCAGAGGGCTCC
Phenotype
MIM: 616793
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G2F PubMed Links

Gene Details

Gene
PLA2G2F
Gene Name
phospholipase A2 group IIF
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022819.3 260 Missense Mutation ATT,GTT I57V NP_073730.3
XM_011541955.1 260 Missense Mutation ATT,GTT I57V XP_011540257.1
XM_011541956.1 260 Missense Mutation ATT,GTT I57V XP_011540258.1

View Full Product Details