Product Details

SNP ID
hCV166879529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:145868346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCAAGGCGAGCATTTTCTCCCC[G/A]AATTGCTAGAACCTCTTCTGAAAGC
Phenotype
MIM: 605987
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ANKRD35 PubMed Links

Gene Details

Gene
ANKRD35
Gene Name
ankyrin repeat domain 35
There are no transcripts associated with this gene.

Gene
PIAS3
Gene Name
protein inhibitor of activated STAT 3
There are no transcripts associated with this gene.

View Full Product Details