Product Details

SNP ID
rs149214676
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:200407168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTAGAGTTAGTGATAAAGGTCAT[C/T]GGCACCCTGCTGCCCGACTTAAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF281 PubMed Links

Gene Details

Gene
ZNF281
Gene Name
zinc finger protein 281
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281293.1 4050 Silent Mutation CCA,CCG P846P NP_001268222.1
NM_001281294.1 4050 Silent Mutation CCA,CCG P810P NP_001268223.1
NM_012482.4 4050 Silent Mutation CCA,CCG P846P NP_036614.1
XM_017000887.1 4050 Silent Mutation CCA,CCG P846P XP_016856376.1
XM_017000888.1 4050 Silent Mutation CCA,CCG P846P XP_016856377.1

View Full Product Details