Product Details

SNP ID
rs149285870
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:196981202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAAAACAAAACAATTTCTTATG[A/C]AGCAATGCTATATTCAGGAAATATC
Phenotype
MIM: 608593
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CFHR5 PubMed Links

Gene Details

Gene
CFHR5
Gene Name
complement factor H related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030787.3 Intron NP_110414.1
XM_011510020.2 Intron XP_011508322.1

View Full Product Details