Product Details

SNP ID
rs149436737
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:156767610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGCCTTGTTCGGTGGAAATCAGC[C/T]GTAGCCATGAGTTTCTGCCGGGGCT
Phenotype
MIM: 179755
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRCC PubMed Links

Gene Details

Gene
PRCC
Gene Name
papillary renal cell carcinoma (translocation-associated)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005973.4 129 UTR 5 NP_005964.3
XM_005245313.2 129 UTR 5 XP_005245370.1
XM_005245314.1 129 UTR 5 XP_005245371.1
XM_011509726.1 129 UTR 5 XP_011508028.1

View Full Product Details