Product Details

SNP ID
rs149753769
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:145871999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCACCTTGGCTTCCTTGTCTC[G/A]GCAAGCCCCAATGAGATGCTCCATC
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ANKRD35 PubMed Links

Gene Details

Gene
ANKRD35
Gene Name
ankyrin repeat domain 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280799.1 2878 Silent Mutation AGA,CGA R834R NP_001267728.1
NM_144698.4 2878 Silent Mutation AGA,CGA R924R NP_653299.4

View Full Product Details