Product Details

SNP ID
rs150621373
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:205599190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGGCGTGATCTTTGGTTGTCT[C/G]GCTTTTACCTTCTATATCTTGCTCC
Phenotype
MIM: 600246
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ELK4 PubMed Links

Gene Details

Gene
ELK4
Gene Name
ELK4, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001973.3 1514 Intron NP_001964.2
NM_021795.2 1514 Intron NP_068567.1
XM_005244950.4 1514 Intron XP_005245007.1
XM_005244951.4 1514 Intron XP_005245008.1
XM_017000580.1 1514 Intron XP_016856069.1
Gene
MFSD4A
Gene Name
major facilitator superfamily domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181644.4 1514 Silent Mutation CTC,CTG L476L NP_857595.3
XM_011509229.2 1514 Silent Mutation CTC,CTG L389L XP_011507531.1

View Full Product Details