Product Details
- SNP ID
-
rs150630761
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:247451254 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGATCATCAGGTGGGAGGAACACGT[C/T]CCAAAGGCCTTGTGTCGTCCCTTGG
- Phenotype
-
MIM: 606416
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NLRP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12086048] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NLRP3
- Gene Name
- NLR family pyrin domain containing 3
There are no transcripts associated with this gene.
- Gene
- OR2B11
- Gene Name
- olfactory receptor family 2 subfamily B member 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004492.1 |
729 |
Silent Mutation |
GGA,GGG |
G243G |
NP_001004492.1 |
View Full Product Details