Product Details

SNP ID
rs150723240
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:16045628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGTACTTCCTGATGACCCTCGG[A/G]GTGCTCATGGCCCTGGTCAGCTGTG
Phenotype
MIM: 602023
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCNKB PubMed Links

Gene Details

Gene
CLCNKB
Gene Name
chloride voltage-gated channel Kb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000085.4 323 Silent Mutation GGA,GGG G57G NP_000076.2
NM_001165945.2 323 Intron NP_001159417.2

View Full Product Details