Product Details

SNP ID
rs150839676
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:11056698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCACCCACATGTAGGGTCAGCTT[C/T]GAGCTAGAGTAGCCAATGGCCATGC
Phenotype
MIM: 605960 MIM: 605102 MIM: 182891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
There are no transcripts associated with this gene.

Gene
MASP2
Gene Name
mannan binding lectin serine peptidase 2
There are no transcripts associated with this gene.

Gene
SRM
Gene Name
spermidine synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003132.2 532 Silent Mutation TCA,TCG S147S NP_003123.2
XM_017002179.1 532 Silent Mutation TCA,TCG S147S XP_016857668.1

View Full Product Details