Product Details

SNP ID
rs151267286
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:154926405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGGCTCTGCTCCAACGCTACAA[C/T]GCGGACCGTCTGCGTCACGGCCCCA
Phenotype
MIM: 607622
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PMVK PubMed Links

Gene Details

Gene
PMVK
Gene Name
phosphomevalonate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323011.1 714 Missense Mutation NP_001309940.1
NM_001323012.1 714 Missense Mutation NP_001309941.1
NM_006556.3 714 Missense Mutation NP_006547.1

View Full Product Details