Product Details

SNP ID
rs151293721
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:200407303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCTGAGATGGAATCTGAAAGCC[C/T]GTTGAAGACTCTAAGTTTTTCTGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF281 PubMed Links

Gene Details

Gene
ZNF281
Gene Name
zinc finger protein 281
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281293.1 3915 Silent Mutation ACA,ACG T801T NP_001268222.1
NM_001281294.1 3915 Silent Mutation ACA,ACG T765T NP_001268223.1
NM_012482.4 3915 Silent Mutation ACA,ACG T801T NP_036614.1
XM_017000887.1 3915 Silent Mutation ACA,ACG T801T XP_016856376.1
XM_017000888.1 3915 Silent Mutation ACA,ACG T801T XP_016856377.1

View Full Product Details