Product Details

SNP ID
rs138162380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63977002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCATGCTGGGAGAAGGCCTCCAC[A/G]TAGACGAGGTAGTTGTGGGGACAGT
Phenotype
MIM: 613979
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRPF6 PubMed Links

Gene Details

Gene
PRPF6
Gene Name
pre-mRNA processing factor 6
There are no transcripts associated with this gene.

Gene
SAMD10
Gene Name
sterile alpha motif domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080621.4 589 Silent Mutation TAC,TAT Y138Y NP_542188.1
XM_005260199.4 589 Silent Mutation TAC,TAT Y177Y XP_005260256.1
XM_006723705.3 589 Silent Mutation TAC,TAT Y144Y XP_006723768.1
XM_011528565.2 589 Silent Mutation TAC,TAT Y342Y XP_011526867.1
XM_017027671.1 589 Silent Mutation TAC,TAT Y138Y XP_016883160.1
Gene
ZNF512B
Gene Name
zinc finger protein 512B
There are no transcripts associated with this gene.

View Full Product Details