Product Details

SNP ID
rs140498723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18289738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAACAGTCTCTGGTGTATACTAA[A/G]TGTTCAGTGAATATTACCTTTCATT
Phenotype
MIM: 604075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 1105 Intron NP_001076799.1
NM_001282995.1 1105 Intron NP_001269924.1
NM_001282996.1 1105 Intron NP_001269925.1
NM_001282997.1 1105 Intron NP_001269926.1
NM_001282998.1 1105 Intron NP_001269927.1
NM_001282999.1 1105 Intron NP_001269928.1
NM_001283000.1 1105 Intron NP_001269929.1
NM_001283001.1 1105 Intron NP_001269930.1
NM_001283002.1 1105 Intron NP_001269931.1
NM_001283003.1 1105 Intron NP_001269932.1
NM_001283004.1 1105 Intron NP_001269933.1
NM_001283005.1 1105 Intron NP_001269934.1
NM_001283006.1 1105 Intron NP_001269935.1
NM_001283007.1 1105 Intron NP_001269936.1
NM_001283008.1 1105 Intron NP_001269937.1
NM_003434.5 1105 Intron NP_003425.2
XM_005260819.1 1105 Intron XP_005260876.1
XM_005260820.4 1105 Intron XP_005260877.1
XM_011529336.1 1105 Intron XP_011527638.1
XM_011529337.1 1105 Intron XP_011527639.1
XM_011529338.2 1105 Intron XP_011527640.1
XM_011529339.1 1105 Intron XP_011527641.1
XM_011529340.1 1105 Intron XP_011527642.1
XM_017028041.1 1105 Intron XP_016883530.1
XM_017028042.1 1105 UTR 5 XP_016883531.1
XM_017028043.1 1105 Intron XP_016883532.1
XM_017028044.1 1105 Intron XP_016883533.1
XM_017028045.1 1105 Intron XP_016883534.1
XM_017028046.1 1105 Intron XP_016883535.1
XM_017028047.1 1105 UTR 5 XP_016883536.1
XM_017028048.1 1105 UTR 5 XP_016883537.1
XM_017028049.1 1105 Intron XP_016883538.1
XM_017028050.1 1105 Intron XP_016883539.1
XM_017028051.1 1105 Intron XP_016883540.1
XM_017028052.1 1105 Intron XP_016883541.1
XM_017028053.1 1105 Intron XP_016883542.1
XM_017028054.1 1105 Intron XP_016883543.1
XM_017028055.1 1105 Intron XP_016883544.1
XM_017028056.1 1105 Intron XP_016883545.1
XM_017028057.1 1105 Intron XP_016883546.1
XM_017028058.1 1105 Intron XP_016883547.1

View Full Product Details