Product Details

SNP ID
rs140764670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44542398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTACTCCGTGTTTCTGCTAATCA[C/T]CAGAATGAAACTTAAAAAAAAAAAG
Phenotype
MIM: 604932
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKIG PubMed Links

Gene Details

Gene
PKIG
Gene Name
protein kinase (cAMP-dependent, catalytic) inhibitor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281444.1 Intron NP_001268373.1
NM_001281445.1 Intron NP_001268374.1
NM_007066.4 Intron NP_008997.1
NM_181804.2 Intron NP_861520.1
NM_181805.2 Intron NP_861521.1
XM_017027614.1 Intron XP_016883103.1
XM_017027615.1 Intron XP_016883104.1
XM_017027616.1 Intron XP_016883105.1

View Full Product Details