Product Details

SNP ID
rs141107623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31406134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGAGCAGAGTAACAGTCAAAA[A/G]CAGAAGAAGGAGCTTCATGAATGAT
Phenotype
MIM: 616075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEFB121 PubMed Links

Gene Details

Gene
DEFB121
Gene Name
defensin beta 121
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011878.2 133 Missense Mutation CTT,TTT L7F NP_001011878.1
NM_001171832.1 133 Intron NP_001165303.1
XM_005260383.3 133 Intron XP_005260440.1

View Full Product Details