Product Details

SNP ID
rs141960100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45538007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTCCACGGCTGAACGGGCAA[C/T]ACTTCATGTTTTCTGGGCAATCTCT
Phenotype
MIM: 609031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPPIN PubMed Links

Gene Details

Gene
EPPIN
Gene Name
epididymal peptidase inhibitor
There are no transcripts associated with this gene.

Gene
EPPIN-WFDC6
Gene Name
EPPIN-WFDC6 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198986.1 548 Missense Mutation TAT,TGT Y160C NP_001185915.1
Gene
WFDC6
Gene Name
WAP four-disulfide core domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080827.1 548 Missense Mutation TAT,TGT Y60C NP_543017.1

View Full Product Details