Product Details

SNP ID
rs142108350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64165602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTTCTCACTGCTCCTCCTGAGT[C/T]ACCAAGGGCTTTGCTTGTCTCAGCA
Phenotype
MIM: 600379
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYT1 PubMed Links

Gene Details

Gene
MYT1
Gene Name
myelin transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004535.2 Intron NP_004526.1

View Full Product Details