Product Details

SNP ID
rs142859533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58997667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTCCCAGAGAGGGAGCCGTAGT[C/T]TCCCACCCTCCAGAGGGTGTAGTTC
Phenotype
MIM: 603169 MIM: 605297
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSZ PubMed Links

Gene Details

Gene
CTSZ
Gene Name
cathepsin Z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001336.3 700 Missense Mutation AAC,GAC N192D NP_001327.2
Gene
NELFCD
Gene Name
negative elongation factor complex member C/D
There are no transcripts associated with this gene.

View Full Product Details