Product Details

SNP ID
rs145012822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3252115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGCAGGGGAGGCAGGCACTCCA[C/T]TGGGGAAGATGGCTCTCGTGGAGCA
Phenotype
MIM: 614146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf194 PubMed Links

Gene Details

Gene
C20orf194
Gene Name
chromosome 20 open reading frame 194
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009984.2 4096 UTR 3 NP_001009984.1
XM_005260684.3 4096 UTR 3 XP_005260741.1
XM_005260687.4 4096 UTR 3 XP_005260744.1
XM_011529208.2 4096 UTR 3 XP_011527510.1
XM_017027783.1 4096 UTR 3 XP_016883272.1

View Full Product Details