Product Details

SNP ID
rs145824632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44755373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCTCCTCCACCGGAACACAGA[C/T]GACAACATTCTTTCTAGTCCGCCAG
Phenotype
MIM: 607368 MIM: 611601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNK15 PubMed Links

Gene Details

Gene
KCNK15
Gene Name
potassium two pore domain channel subfamily K member 15
There are no transcripts associated with this gene.

Gene
KCNK15-AS1
Gene Name
KCNK15 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
RIMS4
Gene Name
regulating synaptic membrane exocytosis 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205317.1 1638 UTR 3 NP_001192246.1
NM_182970.3 1638 UTR 3 NP_892015.1

View Full Product Details