Product Details

SNP ID
rs147350283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38728030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCTACACGTCTCAGATCTTCCT[A/G]CCTTCGCTGGAGGGCAATATGCAGC
Phenotype
MIM: 616440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC32A1 PubMed Links

Gene Details

Gene
SLC32A1
Gene Name
solute carrier family 32 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080552.2 1232 Silent Mutation CTA,CTG L323L NP_542119.1

View Full Product Details