Product Details
- SNP ID
-
rs148802229
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45221750 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGCCAAGCGGATCTTCCCAATTT[A/C]CACATGGACAAAAGGGCCAGCACTA
- Phenotype
-
MIM: 182141
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SEMG2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2233896] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEMG2
- Gene Name
- semenogelin II
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003008.2 |
139 |
Missense Mutation |
ACA,CCA |
T40P |
NP_002999.1 |
View Full Product Details