Product Details

SNP ID
rs149386883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3146956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCCTGGAGCCATAGCAATACTG[A/G]TTCCTGTTTGTGAACTGAACAGCCA
Phenotype
MIM: 614272
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FASTKD5 PubMed Links

Gene Details

Gene
FASTKD5
Gene Name
FAST kinase domains 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021826.4 2426 Silent Mutation AAC,AAT N705N NP_068598.1
Gene
UBOX5
Gene Name
U-box domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267584.1 2426 Intron NP_001254513.1
NM_014948.3 2426 Intron NP_055763.1
NM_199415.2 2426 Intron NP_955447.1
Gene
UBOX5-AS1
Gene Name
UBOX5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details