Product Details
- SNP ID
-
rs149460018
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:2484055 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGACTTCTGGCTAAAACCTCGCCC[A/G]CACTCACTGCACACATAAGGCTTCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF343
PubMed Links
Gene Details
- Gene
- ZNF343
- Gene Name
- zinc finger protein 343
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282495.1 |
1258 |
Silent Mutation |
TGC,TGT |
C302C |
NP_001269424.1 |
NM_001282496.1 |
1258 |
Silent Mutation |
TGC,TGT |
C302C |
NP_001269425.1 |
NM_001282497.1 |
1258 |
Silent Mutation |
TGC,TGT |
C343C |
NP_001269426.1 |
NM_001282498.1 |
1258 |
Silent Mutation |
TGC,TGT |
C212C |
NP_001269427.1 |
NM_001282499.1 |
1258 |
Intron |
|
|
NP_001269428.1 |
NM_001321800.1 |
1258 |
Silent Mutation |
TGC,TGT |
C302C |
NP_001308729.1 |
NM_001321801.1 |
1258 |
Silent Mutation |
TGC,TGT |
C343C |
NP_001308730.1 |
NM_001321802.1 |
1258 |
Silent Mutation |
TGC,TGT |
C342C |
NP_001308731.1 |
NM_001321803.1 |
1258 |
Silent Mutation |
TGC,TGT |
C342C |
NP_001308732.1 |
NM_001321805.1 |
1258 |
Silent Mutation |
TGC,TGT |
C301C |
NP_001308734.1 |
NM_024325.5 |
1258 |
Silent Mutation |
TGC,TGT |
C302C |
NP_077301.4 |
XM_017028062.1 |
1258 |
Silent Mutation |
TGC,TGT |
C302C |
XP_016883551.1 |
View Full Product Details