Product Details

SNP ID
rs150135017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3249437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGCTACTTTTATATATTTACT[A/G]AATTAAGTAGTGTAATAAATACTAT
Phenotype
MIM: 614146 MIM: 610206
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf194 PubMed Links

Gene Details

Gene
C20orf194
Gene Name
chromosome 20 open reading frame 194
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009984.2 6774 UTR 3 NP_001009984.1
XM_005260684.3 6774 Intron XP_005260741.1
XM_005260687.4 6774 Intron XP_005260744.1
XM_011529208.2 6774 Intron XP_011527510.1
XM_017027783.1 6774 Intron XP_016883272.1
Gene
SLC4A11
Gene Name
solute carrier family 4 member 11
There are no transcripts associated with this gene.

View Full Product Details