Product Details
- SNP ID
-
rs151101129
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63975750 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGCACCTGCAGGCGGAGCACCTG[C/T]TGCAGCACCTCCTGCCTCTGGGCCT
- Phenotype
-
MIM: 613979
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRPF6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs817343] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRPF6
- Gene Name
- pre-mRNA processing factor 6
There are no transcripts associated with this gene.
- Gene
- SAMD10
- Gene Name
- sterile alpha motif domain containing 10
- Gene
- ZNF512B
- Gene Name
- zinc finger protein 512B
There are no transcripts associated with this gene.
View Full Product Details