Product Details

SNP ID
rs149982226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25583168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTTCCAGATTTCACACAGTTA[C/T]GTCTAATTGGTGAGCCCATGCTACC
Phenotype
MIM: 609337 MIM: 611845
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00515 PubMed Links

Gene Details

Gene
LINC00515
Gene Name
long intergenic non-protein coding RNA 515
There are no transcripts associated with this gene.

Gene
MIR155
Gene Name
microRNA 155
There are no transcripts associated with this gene.

Gene
MIR155HG
Gene Name
MIR155 host gene
There are no transcripts associated with this gene.

Gene
MRPL39
Gene Name
mitochondrial ribosomal protein L39
There are no transcripts associated with this gene.

View Full Product Details